Oxaleddikesyre ≥97.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
O0072-5G
O0072-25G
TCIAO0072-5GEA
255
DKK
TCIAO0072-5G
TCIAO0072-25G
Oxaleddikesyre ≥97.0% (by titrimetric analysis)
Oxaleddikesyre
Formel:
C₄H₄O₅ Kogepunkt: 310 °C (1013 hPa) Smeltepunkt: 182…183 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00002592 CAS-nummer: 328-42-7 EINECS: 206-329-8 UN nummer: 3261 ADR: 8,II Merck Index: 14,06909 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | min. 97.0 % |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...