Ethylencarbonat ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1,3-Dioxa-2-cyclopentanone

E0076-25G E0076-500G
TCIAE0076-25GEA 140 DKK
TCIAE0076-25G TCIAE0076-500G
Ethylencarbonat ≥99.0% (by GC)
Ethylencarbonat
Formel: C₃H₄O₃
Kogepunkt: 243 °C (1013 hPa)
Smeltepunkt: 36 °C
Massefylde: 1,47 g/cm³ (20 °C)
Flammepunkt: 150 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00005382
CAS-nummer: 96-49-1
EINECS: 202-510-0

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 99.0 %
Melting point 37.0 to 39.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR