4-Nitrophthalonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Nitrobenzene-1,2-dicarbonitrile, 4-Nitro-1,2-benzenedicarbonitrile, 4-Nitro-1,2-dicyanobenzene

N0524-25G N0524-100G
TCIAN0524-25GEA 386 DKK
TCIAN0524-25G TCIAN0524-100G
4-Nitrophthalonitril ≥98.0% (by GC)
4-Nitrophthalonitril
Formel: C₈H₃N₃O₂
Smeltepunkt: 144 °C
MDL Number: MFCD00040301
CAS-nummer: 31643-49-9
EINECS: 250-748-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc98fcee46312eaa3b226d60d1b92f25997f5e31&vid=cc98fcee46312eaa3b226d60d1b92f25997f5e31&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 142.0 to 145.0 deg-C
Solubility in Acetone almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR