Kobber(I)cyanid ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Copper cyanide, Cuprous cyanide

C1952-300G C1952-25G
TCIAC1952-300GEA 247 DKK
TCIAC1952-300G TCIAC1952-25G
Kobber(I)cyanid ≥98.0% (by titrimetric analysis)
Kobber(I)cyanid
Formel: CCuN
Molekylvægt: 89.56 g/mol
Smeltepunkt: 473°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010975
CAS-nummer: 544-92-3
EINECS: 208-883-6
UN nummer: 1587
ADR: 6.1 II
Merck Index: 14,02661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43ce64b7b666092aa42260ad1ce8618b21bb6500&vid=43ce64b7b666092aa42260ad1ce8618b21bb6500&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Dark green powder to crystaline
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR