3-Bromthioanisol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-Bromophenyl methyl sulphide, 1-Bromo-3-(methylthio)benzene
B2346-25G
B2346-5G
TCIAB2346-25GEA
1800
DKK
TCIAB2346-25G
TCIAB2346-5G
3-Bromthioanisol ≥98.0% (by GC)
3-Bromthioanisol
Formel:
C₇H₇BrS Molekylvægt: 203.1 g/mol Kogepunkt: 250°C |
MDL Number:
MFCD00041395 CAS-nummer: 33733-73-2 |
Specifikationer Testresultater
Appearance | Light orange to Yellow to Green clear liquid to slightly cloudy liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...