α-Brom-2-(trifluormethoxy)toluen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

T2311-1G T2311-5G
TCIAT2311-1GEA 304 DKK
TCIAT2311-1G TCIAT2311-5G
α-Brom-2-(trifluormethoxy)toluen ≥98.0% (by GC)
α-Brom-2-(trifluormethoxy)toluen
Formel: C₈H₆BrF₃O
Molekylvægt: 255.03 g/mol
Kogepunkt: 222
Smeltepunkt: 32
Massefylde: 1.583
MDL Number: MFCD00236321
CAS-nummer: 198649-68-2
UN nummer: 3265
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6ec603794ebd6984a50641d184031b508f105c1&vid=d6ec603794ebd6984a50641d184031b508f105c1&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR