5-Brom-1-indanon ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-Bromo-2,3-dihydro-1H-inden-1-one

B2342-1G B2342-25G B2342-5G
TCIAB2342-1GEA 189 DKK
TCIAB2342-1G TCIAB2342-25G TCIAB2342-5G
5-Brom-1-indanon ≥98.0% (by GC)
5-Brom-1-indanon
Formel: C₉H₇BrO
Kogepunkt: 280 °C (1013 hPa)
Smeltepunkt: 126…127 °C
MDL Number: MFCD00082718
CAS-nummer: 34598-49-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d06c1e77687edccc75147cde3f3b3ba8bceb238&vid=4d06c1e77687edccc75147cde3f3b3ba8bceb238&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 124.0 to 129.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR