2'-Bromacetophenon ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B0538-25G
B0538-5G
TCIAB0538-25GEA
829
DKK
TCIAB0538-25G
TCIAB0538-5G
2'-Bromacetophenon ≥98.0% (by GC)
2'-Bromacetophenon
Formel:
C₈H₇BrO Kogepunkt: 136°C Smeltepunkt: 0 °C Massefylde: 1,476 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000067 CAS-nummer: 2142-69-0 EINECS: 218-398-1 |
|
Specifikationer Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...