11-Brom-1-undecen ≥94.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B3576-5G B3576-25G
TCIAB3576-5GEA 444 DKK
TCIAB3576-5G TCIAB3576-25G
11-Brom-1-undecen ≥94.0% (by GC)
11-Bromo-1-undecene
Formel: C₁₁H₂₁Br
Molekylvægt: 233.19 g/mol
Kogepunkt: 115°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00040825
CAS-nummer: 7766-50-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f64b36e21a91e59c68439e5bcfda69f7e761bc9&vid=6f64b36e21a91e59c68439e5bcfda69f7e761bc9&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR