2-(2-Pyridyl)pyridin ≥99.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: α,α′-Dipyridyl, 2,2'-Bipyridine, 2,2'-Dipyridyl

B0468-25G B0468-500G B0468-100G
TCIAB0468-25GEA 321 DKK
TCIAB0468-25G TCIAB0468-500G TCIAB0468-100G
2-(2-Pyridyl)pyridin ≥99.0% (by titrimetric analysis)
2,2'-Bipyridyl
Formel: C₁₀H₈N₂
Kogepunkt: 273°C
Smeltepunkt: 70…72 °C
Massefylde: 1,326 g/cm³ (-150 °C)
Flammepunkt: 121 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006212
CAS-nummer: 366-18-7
EINECS: 206-674-4
UN nummer: 2811
ADR: 6.1,III
Merck Index: 14,03347
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f034e3d688e952a90bfb14613620ae5c61137e11&vid=f034e3d688e952a90bfb14613620ae5c61137e11&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 69.5 to 72.0 deg-C
Solubility in EtOH transparency
Ignition residue (Sulfate) max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR