Pyrazin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
P0544-25G
P0544-500G
TCIAP0544-25GEA
222
DKK
TCIAP0544-25G
TCIAP0544-500G
Pyrazin ≥98.0% (by GC)
Pyrazine
Formel:
C₄H₄N₂ Molekylvægt: 80.09 g/mol Kogepunkt: 115 Smeltepunkt: 52 |
MDL Number:
MFCD00006122 CAS-nummer: 290-37-9 EINECS: 206-027-6 UN nummer: 1325 ADR: 4.1 III Merck Index: 14,07957 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 52.0 to 55.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...