Phenazin ≥99.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
P0082-1G
P0082-25G
P0082-5G
TCIAP0082-1GEA
132
DKK
TCIAP0082-1G
TCIAP0082-25G
TCIAP0082-5G
Phenazin ≥99.0% (by HPLC)
Phenazine
Formel:
C₁₂H₈N₂ Molekylvægt: 180.21 g/mol Kogepunkt: 339 |
MDL Number:
MFCD00005023 CAS-nummer: 92-82-0 EINECS: 202-193-9 Merck Index: 14,07217 |
|
Specifikationer Testresultater
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 174.0 to 178.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...