1-Ethylpiperidin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

E0166-25ML E0166-500ML
TCIAE0166-25MLEA 312 DKK
TCIAE0166-25ML TCIAE0166-500ML
1-Ethylpiperidin ≥98.0% (by GC, titration analysis)
1-Ethylpiperidin
Formel: C₇H₁₅N
Kogepunkt: 131°C
Smeltepunkt: –57 °C
Massefylde: 0,824 g/cm³ (20 °C)
Flammepunkt: 19°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006507
CAS-nummer: 766-09-6
EINECS: 212-161-6
UN nummer: 2386
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef0cdb5983294079ad0cd8c79ce311dd352b7dd9&vid=ef0cdb5983294079ad0cd8c79ce311dd352b7dd9&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR