Oxacyclohexan ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

T0110-25ML T0110-500ML T0110-100ML
TCIAT0110-25MLEA 132 DKK
TCIAT0110-25ML TCIAT0110-500ML TCIAT0110-100ML
Oxacyclohexan ≥98.0% (by GC)
Tetrahydropyran
Formel: C₅H₁₀O
Molekylvægt: 86.13 g/mol
Kogepunkt: 88°C
Smeltepunkt: -44.2
Flammepunkt: –20°C
MDL Number: MFCD00006585
CAS-nummer: 142-68-7
EINECS: 205-552-8
UN nummer: 1993
ADR: 3 II
Merck Index: 14,09217
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea5ab73fbe62acf59ab28320028c4ae0f55fd68&vid=5ea5ab73fbe62acf59ab28320028c4ae0f55fd68&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR