Benzensulfonylchlorid ≥99.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
B0036-25G
B0036-500G
TCIAB0036-25GEA
132
DKK
TCIAB0036-25G
TCIAB0036-500G
Benzensulfonylchlorid ≥99.0% (by GC, titration analysis)
Benzensulfonylchlorid
Formel:
C₆H₅ClO₂S Kogepunkt: 252°C Smeltepunkt: 15 °C Massefylde: 1,3842 g/cm³ (20 °C) Flammepunkt: 132°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007426 CAS-nummer: 98-09-9 EINECS: 202-636-6 UN nummer: 2225 ADR: 8,III Merck Index: 14,01072 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 99.0 % |
Purity (Neutralization titration) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...