1,4-Diphenoxybenzen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
D2017-25G
D2017-5G
TCIAD2017-25GEA
936
DKK
TCIAD2017-25G
TCIAD2017-5G
1,4-Diphenoxybenzen ≥98.0% (by GC)
1,4-Diphenoxybenzen
Formel:
C₁₈H₁₄O₂ Kogepunkt: 182°C |
MDL Number:
MFCD00038368 CAS-nummer: 3061-36-7 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 75.0 to 78.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...