Pyrogallol ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
1,2,3-Trihydroxybenzen
P0570-25G
P0570-500G
P0570-100G
TCIAP0570-25GEA
165
DKK
TCIAP0570-25G
TCIAP0570-500G
TCIAP0570-100G
Pyrogallol ≥99.0% (by GC)
Pyrogallol
Formel:
C₆H₆O₃ Molekylvægt: 126.11 g/mol Kogepunkt: 309°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002192 CAS-nummer: 87-66-1 EINECS: 201-762-9 Merck Index: 14,08000 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 130.0 to 136.0 deg-C |
Solubility in Water | transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...