Tetrabutoxysilan ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Tetrabutyl orthosilicate

T1126-25ML T1126-500ML
TCIAT1126-25MLEA 156 DKK
TCIAT1126-25ML TCIAT1126-500ML
Tetrabutoxysilan ≥98.0% (by GC)
Tetrabutoxysilane
Formel: C₁₆H₃₆O₄Si
Molekylvægt: 320.54 g/mol
Kogepunkt: 277°C
Flammepunkt: 136°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009432
CAS-nummer: 4766-57-8
EINECS: 225-305-8
UN nummer: 1760
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=172d03f06940789cb5228ffcdecd231cf73e7a29&vid=172d03f06940789cb5228ffcdecd231cf73e7a29&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR