Bis(2-butoxyethyl)ether ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Diethylene glycol dibutyl ether, 2,2-Dibutoxyethyl ether
B0828-25ML
B0828-500ML
TCIAB0828-25MLEA
132
DKK
TCIAB0828-25ML
TCIAB0828-500ML
Bis(2-butoxyethyl)ether ≥98.0% (by GC)
Bis(2-butoxyethyl)ether
Formel:
C₁₂H₂₆O₃ Molekylvægt: 218.34 g/mol Kogepunkt: 255°C Smeltepunkt: –60°C Flammepunkt: 122°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009459 CAS-nummer: 112-73-2 EINECS: 204-001-9 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...