Pyridin-1-oxid ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

P0557-25G P0557-500G P0557-100G
TCIAP0557-25GEA 181 DKK
TCIAP0557-25G TCIAP0557-500G TCIAP0557-100G
Pyridin-1-oxid ≥98.0% (by titrimetric analysis)
Pyridine-N-oxide
Formel: C₅H₅NO
Kogepunkt: 270 °C (1013 hPa)
Smeltepunkt: 61…64 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00006194
CAS-nummer: 694-59-7
EINECS: 211-774-6
Merck Index: 14,07972
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcc00bd76771a70b49cef7404dd78b68ce05ca6c&vid=bcc00bd76771a70b49cef7404dd78b68ce05ca6c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 62.0 to 67.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR