4-Nitrothioanisol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Nitrophenyl methyl sulphide, 1-Nitro-4-(methylthio)benzene
N0761-25G
N0761-5G
TCIAN0761-25GEA
1620
DKK
TCIAN0761-25G
TCIAN0761-5G
4-Nitrothioanisol ≥98.0% (by GC)
4-Nitrothioanisol
Formel:
C₇H₇NO₂S Kogepunkt: 140°C Smeltepunkt: 66…68 °C |
MDL Number:
MFCD00010868 CAS-nummer: 701-57-5 |
|
Specifikationer Testresultater
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 69.0 to 73.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...