2-Methoxy-3-nitropyridin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

M1395-25G M1395-5G
TCIAM1395-25GEA 2290 DKK
TCIAM1395-25G TCIAM1395-5G
2-Methoxy-3-nitropyridin ≥98.0% (by GC)
2-Methoxy-3-nitropyridine
Formel: C₆H₆N₂O₃
Smeltepunkt: 149…150 °C
MDL Number: MFCD00023459
CAS-nummer: 20265-35-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a19a3367e253e19f31cadb2bff6c06f5b1b550a2&vid=a19a3367e253e19f31cadb2bff6c06f5b1b550a2&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Amber powder to crystal
Purity (GC) min. 98.0 %
Melting point 56.0 to 58.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR