Dicyclohexylamin ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
DCHA
D0435-25ML
D0435-500ML
TCIAD0435-25MLEA
132
DKK
TCIAD0435-25ML
TCIAD0435-500ML
Dicyclohexylamin ≥99.0% (by GC)
Dicyclohexylamin
Formel:
C₁₂H₂₃N Kogepunkt: 256°C Smeltepunkt: 0°C Massefylde: 0,91 g/cm³ (20 °C) Flammepunkt: 105°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011658 CAS-nummer: 101-83-7 EINECS: 202-980-7 UN nummer: 2565 ADR: 8,III Merck Index: 14,03095 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 99.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...