N-Isopropylethylendiamin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: N-(2-Aminoethyl)-N-isopropylamine, N1-Isopropylethane-1,2-diamine

I0381-10ML I0381-25ML
TCIAI0381-10MLEA 517 DKK
TCIAI0381-10ML TCIAI0381-25ML
N-Isopropylethylendiamin ≥98.0% (by GC, titration analysis)
N-Isopropylethylendiamin
Formel: C₅H₁₄N₂
Molekylvægt: 102.18 g/mol
Kogepunkt: 138°C
Flammepunkt: 6°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008164
CAS-nummer: 19522-67-9
EINECS: 243-125-8
UN nummer: 2733
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d252424cf5cbc8e8aa6ce7f3be1ceef7ed630e0&vid=9d252424cf5cbc8e8aa6ce7f3be1ceef7ed630e0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR