Guanin ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Amino-1H-purin-6(7H)-one

G0169-1G G0169-25G G0169-250G
TCIAG0169-1GEA 132 DKK
TCIAG0169-1G TCIAG0169-25G TCIAG0169-250G
Guanin ≥98.0% (by HPLC, titration analysis)
Guanin
Formel: C₅H₅N₅O
Molekylvægt: 151.13 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00071533
CAS-nummer: 73-40-5
EINECS: 200-799-8
Merck Index: 14,04564
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fae9141643408e2e2090c932a85d1a47f788636&vid=6fae9141643408e2e2090c932a85d1a47f788636&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in hot dilute HCl almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR