o-Anisidin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Methoxyanilin, 2-Anisidin

A0486-25ML A0486-500ML
TCIAA0486-25MLEA 132 DKK
TCIAA0486-25ML TCIAA0486-500ML
o-Anisidin ≥98.0% (by GC, titration analysis)
o-Anisidin
Formel: C₇H₉NO
Molekylvægt: 123.15 g/mol
Kogepunkt: 225°C
Smeltepunkt: 5
Flammepunkt: 111°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00007688
CAS-nummer: 90-04-0
EINECS: 201-963-1
UN nummer: 2431
ADR: 6.1 III
Merck Index: 14,00667
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b13c2594ec56be0a04ee9e9015373cb8e3af5ed&vid=2b13c2594ec56be0a04ee9e9015373cb8e3af5ed&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR