o-Toluidin ≥99.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Toluidin

T0299-25G T0299-500G
TCIAT0299-25GEA 132 DKK
TCIAT0299-25G TCIAT0299-500G
o-Toluidin ≥99.0% (by GC, titration analysis)
o-Toluidin
Formel: C₇H₉N
Molekylvægt: 107.16 g/mol
Kogepunkt: 201°C
Flammepunkt: 86°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007730
CAS-nummer: 95-53-4
EINECS: 202-429-0
UN nummer: 1708
ADR: 6.1 II
Merck Index: 14,09536
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=288ed467b55eff8b79e9f1cf86cb44987ce6df25&vid=288ed467b55eff8b79e9f1cf86cb44987ce6df25&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR