2-(Dimethylamino)pyridin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N,N-Dimethylpyridin-2-amine

D1680-25G D1680-500G
TCIAD1680-25GEA 575 DKK
TCIAD1680-25G TCIAD1680-500G
2-(Dimethylamino)pyridin ≥98.0% (by GC, titration analysis)
2-(Dimethylamino)pyridine
Formel: C₇H₁₀N₂
Molekylvægt: 122.17 g/mol
Kogepunkt: 108°C
Flammepunkt: 74°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006261
CAS-nummer: 5683-33-0
EINECS: 227-147-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a283b089b14ed77f56d57fc2da709353bd78e46b&vid=a283b089b14ed77f56d57fc2da709353bd78e46b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR