N-Iodsuccinimid ≥99.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
I0074-100G
I0074-25G
I0074-5G
TCIAI0074-100GEA
2200
DKK
TCIAI0074-100G
TCIAI0074-25G
TCIAI0074-5G
N-Iodsuccinimid ≥99.0% (by titrimetric analysis)
N-Iodsuccinimid
Formel:
C₄H₄INO₂ Kogepunkt: 356 °C (1013 hPa) Smeltepunkt: 200…202 °C Massefylde: 2,245 g/cm³ (20 °C) Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00005512 CAS-nummer: 516-12-1 EINECS: 208-221-6 Merck Index: 14,05045 |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Iodometric Titration) | min. 99.0 % |
Solubility in Acetone | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...