Guanidiniumcarbonat ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Guanidine carbonate salt, Diguanidinium carbonate, Guanidine carbonate

G0161-25G G0161-500G
TCIAG0161-25GEA 132 DKK
TCIAG0161-25G TCIAG0161-500G
Guanidiniumcarbonat ≥98.0% (by titrimetric analysis)
Guanidiniumcarbonat
Formel: C₃H₁₂N₆O₃
Smeltepunkt: 197 °C
Massefylde: 1,251 g/cm³ (20 °C)
MDL Number: MFCD00013029
CAS-nummer: 593-85-1
EINECS: 209-813-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=762603d83a7ba84ff4703e2bbf4be2b756326183&vid=762603d83a7ba84ff4703e2bbf4be2b756326183&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR