2-Fluorthioanisol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Fluoro-2-(methylthio)benzene, 2-Fluorophenyl methyl sulphide

F0565-25G F0565-5G
TCIAF0565-25GEA 2030 DKK
TCIAF0565-25G TCIAF0565-5G
2-Fluorthioanisol ≥98.0% (by GC)
2-Fluorthioanisol
Formel: C₇H₇FS
Kogepunkt: 194°C
Flammepunkt: 91°C
MDL Number: MFCD03093762
CAS-nummer: 655-20-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f586297805c0c6f93d157b44a52f02fc496bd68&vid=0f586297805c0c6f93d157b44a52f02fc496bd68&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR