(4-Hydroxy-3-nitrophenyl)eddikesyre ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-(4-Hydroxy-3-nitrophenyl)acetic acid

H1362-1G H1362-5G
TCIAH1362-1GEA 312 DKK
TCIAH1362-1G TCIAH1362-5G
(4-Hydroxy-3-nitrophenyl)eddikesyre ≥98.0% (by titrimetric analysis)
(4-Hydroxy-3-nitrophenyl)eddikesyre
Formel: (C₆H₃(OH)(NO₂))CH₂COOH
Smeltepunkt: 149…150 °C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00007122
CAS-nummer: 10463-20-4
EINECS: 233-948-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84982fe01b455e2e29ace370a79a662c9db38e29&vid=84982fe01b455e2e29ace370a79a662c9db38e29&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 144.0 to 148.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR