Kobber(II)acetat monohydrat 95,0 Titrimetric
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Copper diacetate monohydrate, Cupric acetate monohydrate, Acetic acid copper (II) salt monohydrate, Copper(II) acetate hydrate
C2346-25G
C2346-500G
TCIAC2346-25GEA
132
DKK
TCIAC2346-25G
TCIAC2346-500G
Kobber(II)acetat monohydrat 95,0 Titrimetric
Kobber(II)acetat monohydrat
Formel:
(H₃CCOO)₂Cu·H₂O Molekylvægt: 199.65 g/mol Smeltepunkt: 115°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00149570 CAS-nummer: 6046-93-1 EINECS: 205-553-3 UN nummer: 3077 ADR: 9 III Merck Index: 13,02651 |
|
Specifikationer Testresultater
Appearance | Blue to Dark blue powder to crystal |
Purity (Chelometric Titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...