trans-Cinnamic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Cinnamic acid (only trans), (2E)-2-Phenyl-2-propenoic acid, trans-Cinnamic acid

C0353-25G C0353-500G C0353-100G
TCIAC0353-25GEA 132 DKK
TCIAC0353-25G TCIAC0353-500G TCIAC0353-100G
trans-Cinnamic acid ≥98.0% (by GC, titration analysis)
trans-Cinnamic acid
Formel: C₉H₈O₂
Kogepunkt: 300°C
Smeltepunkt: 131…134 °C
Massefylde: 1,25 g/cm³ (4 °C)
MDL Number: MFCD00004369
CAS-nummer: 140-10-3
EINECS: 205-398-1
Merck Index: 14,02299
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99fc10c5f81fc4aa86576230911ddc9362fc1bba&vid=99fc10c5f81fc4aa86576230911ddc9362fc1bba&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 132.0 to 136.0 deg-C
Solubility in EtOH very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR