Benzylbutylphthalat ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

P0288-25G P0288-500G
TCIAP0288-25GEA 140 DKK
TCIAP0288-25G TCIAP0288-500G
Benzylbutylphthalat ≥97.0% (by GC)
Benzylbutylphthalat
Formel: C₁₉H₂₀O₄
Kogepunkt: 250°C
Smeltepunkt: –35 °C
Massefylde: 1,12 g/cm³ (20 °C)
Flammepunkt: 213°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009440
CAS-nummer: 85-68-7
EINECS: 201-622-7
UN nummer: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60c98e024878c703cdb20d04bfcc857798390185&vid=60c98e024878c703cdb20d04bfcc857798390185&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR