N,N-Diethylacetamid ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
D0461-25ML
D0461-500ML
TCIAD0461-25MLEA
247
DKK
TCIAD0461-25ML
TCIAD0461-500ML
N,N-Diethylacetamid ≥99.0% (by GC)
N,N-Diethylacetamid
Formel:
C₆H₁₃NO Kogepunkt: 185°C Massefylde: 1,02 g/cm³ (20 °C) Flammepunkt: 78°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009047 CAS-nummer: 685-91-6 EINECS: 211-685-2 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...