Butyroylchlorid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

B0770-25G B0770-500G
TCIAB0770-25GEA 132 DKK
TCIAB0770-25G TCIAB0770-500G
Butyroylchlorid ≥98.0% (by GC, titration analysis)
Butyroylchlorid
Formel: C₄H₇ClO
Kogepunkt: 102°C
Smeltepunkt: –89 °C
Massefylde: 1,0277 g/cm³ (20 °C)
Flammepunkt: 24°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000752
CAS-nummer: 141-75-3
EINECS: 205-498-5
UN nummer: 2353
ADR: 3,II
Merck Index: 14,01598
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=940bf349ff61c9120793744ecd0717e4cf4c2197&vid=940bf349ff61c9120793744ecd0717e4cf4c2197&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) 98.0 to 102.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR