Barbitursyre ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N,N'-Malonylurea, 2,4,6-Trihydroxypyrimidine

B0003-500G B0003-25G
TCIAB0003-500GEA 698 DKK
TCIAB0003-500G TCIAB0003-25G
Barbitursyre ≥98.0% (by titrimetric analysis)
Barbitursyre
Formel: C₄H₄N₂O₃
Molekylvægt: 128.09 g/mol
Kogepunkt: 441
Smeltepunkt: 248°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006666
CAS-nummer: 67-52-7
EINECS: 200-658-0
Merck Index: 14,00963
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc13d578e5c5acd7fde3aa6a862933572f51bd95&vid=bc13d578e5c5acd7fde3aa6a862933572f51bd95&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR