1-Amino-9,10-anthraquinon 98,0 N_ASSAY_METHOD

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Aminoanthraquinone, 1-Aminoanthracene-9,10-dione

A0590-25G A0590-500G
TCIAA0590-25GEA 206 DKK
TCIAA0590-25G TCIAA0590-500G
1-Amino-9,10-anthraquinon 98,0 N_ASSAY_METHOD
1-Amino-9,10-anthraquinon
Formel: C₁₄H₉NO₂
Molekylvægt: 223.23 g/mol
Kogepunkt: 465
MDL Number: MFCD00001213
CAS-nummer: 82-45-1
EINECS: 201-423-5
UN nummer: 3077
ADR: 9 III
Merck Index: 14,00417
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edcae4745851ae26ad3e93fcd6a410f78752977b&vid=edcae4745851ae26ad3e93fcd6a410f78752977b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Orange to Brown to Dark red powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Solubility in hot Acetic acid almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR