9,10-Phenanthrenquinon ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Phenanthrene-9,10-dione
P1136-25G
P1136-250G
TCIAP1136-25GEA
435
DKK
TCIAP1136-25G
TCIAP1136-250G
9,10-Phenanthrenquinon ≥98.0% (by GC)
9,10-Phenanthrenquinon
Formel:
C₁₄H₈O₂ Molekylvægt: 208.22 g/mol Kogepunkt: 360 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001163 CAS-nummer: 84-11-7 EINECS: 201-515-5 UN nummer: 3077 ADR: 9 III Merck Index: 14,07213 |
|
Specifikationer Testresultater
Appearance | Orange to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 207.0 to 211.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...