Methyl-p-benzochinon ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
T1244-25G
T1244-500G
TCIAT1244-25GEA
329
DKK
TCIAT1244-25G
TCIAT1244-500G
Methyl-p-benzochinon ≥98.0% (by GC, titration analysis)
Methyl-p-benzochinon
Formel:
C₇H₆O₂ Kogepunkt: 187 °C (1013 hPa) Smeltepunkt: 65…67 °C Massefylde: 1,08 g/cm³ (75.5 °C) |
MDL Number:
MFCD00001603 CAS-nummer: 553-97-9 EINECS: 209-056-2 |
|
Specifikationer Testresultater
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Iodometric Titration) | min. 98.0 % |
Melting point | 66.0 to 69.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...