1,4-Naphthochinon ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
1,4-Naphthalenedione, 1,4-Dihydro-1,4-diketonaphthalene
N0040-500G
N0040-25G
N0040-100G
TCIAN0040-500GEA
1100
DKK
TCIAN0040-500G
TCIAN0040-25G
TCIAN0040-100G
1,4-Naphthochinon ≥98.0% (by GC)
1,4-Naphthochinon
Formel:
C₁₀H₆O₂ Molekylvægt: 158.16 g/mol Kogepunkt: 298 Smeltepunkt: 124 Flammepunkt: 141 |
MDL Number:
MFCD00001676 CAS-nummer: 130-15-4 EINECS: 204-977-6 UN nummer: 2811 ADR: 6.1 I Merck Index: 12,06482 |
|
Specifikationer Testresultater
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Cerium redox Method) | min. 95.0 % |
Melting point | 123.0 to 126.0 deg-C |
Solubility in hot Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...