4,4'-Bis(diethylamino)benzophenon ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N,N,N',N'-Tetraethyl-4,4'-diaminobenzophenone, 4,4-Bis(diethylamino) benzophenone

B0139-25G B0139-500G B0139-100G
TCIAB0139-25GEA 238 DKK
TCIAB0139-25G TCIAB0139-500G TCIAB0139-100G
4,4'-Bis(diethylamino)benzophenon ≥98.0% (by titrimetric analysis)
4,4'-Bis(diethylamino)benzophenon
Formel: C₂₁H₂₈N₂O
Molekylvægt: 324.47 g/mol
Kogepunkt: 420
Smeltepunkt: 96.5
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009044
CAS-nummer: 90-93-7
EINECS: 202-025-4
UN nummer: 3077
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c55c245653fd9bc16a4c7a38e7f5dd0ce0d91ae4&vid=c55c245653fd9bc16a4c7a38e7f5dd0ce0d91ae4&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 92.0 to 97.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR