o-Acetoxybenzoylchlorid ≥97.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-(Chlorocarbonyl)phenyl acetate, O-Acetylsalicyloyl chloride, 2-Acetylsalicyloyl chloride, 2-Acetoxybenzoyl chloride

A1917-25G A1917-5G
TCIAA1917-25GEA 632 DKK
TCIAA1917-25G TCIAA1917-5G
o-Acetoxybenzoylchlorid ≥97.0% (by HPLC, titration analysis)
o-Acetoxybenzoylchlorid
Formel: C₉H₇ClO₃
Kogepunkt: 135°C
Smeltepunkt: 45…48 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000663
CAS-nummer: 5538-51-2
EINECS: 226-899-1
UN nummer: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c62b5601f27e7a1e0135c1550a577f641f0c170d&vid=c62b5601f27e7a1e0135c1550a577f641f0c170d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to lump
Purity (HPLC) min. 97.0 area%
Purity (Argentmetric Titration) min. 97.0 %
Melting point 48.0 to 52.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR