Propylencarbonat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Propylene glycol carbonate, 4-Methyl-1,3-dioxolan-2-one, Carbonic acid propylene glycol ester

P0525-25G P0525-500G
TCIAP0525-25GEA 140 DKK
TCIAP0525-25G TCIAP0525-500G
Propylencarbonat ≥98.0% (by GC)
Propylencarbonat
Formel: C₄H₆O₃
Molekylvægt: 102.09 g/mol
Kogepunkt: 242°C
Smeltepunkt: –49°C
Flammepunkt: 132°C (closed cup)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005385
CAS-nummer: 108-32-7
EINECS: 203-572-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd4f74a96ae6d9c8ed1a11d6e5993155eefbc650&vid=dd4f74a96ae6d9c8ed1a11d6e5993155eefbc650&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR