3-Methoxypropiononitril ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
M0653-25ML
M0653-500ML
TCIAM0653-25MLEA
132
DKK
TCIAM0653-25ML
TCIAM0653-500ML
3-Methoxypropiononitril ≥99.0% (by GC)
3-Methoxypropiononitril
Formel:
C₄H₇NO Kogepunkt: 166°C Smeltepunkt: -63°C Massefylde: 0,94 g/cm³ (20 °C) Flammepunkt: 65°C |
MDL Number:
MFCD00001958 CAS-nummer: 110-67-8 EINECS: 203-790-7 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...