Cyanoguanidin ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Dicyanodiamide, 1-Cyanoguanidine, N-Cyanoguanidine, Dicyandiamide
C0454-500G
C0454-25G
TCIAC0454-500GEA
230
DKK
TCIAC0454-500G
TCIAC0454-25G
Cyanoguanidin ≥98.0% (by titrimetric analysis)
Cyanoguanidin
Formel:
(H₂N)₂CNCN Kogepunkt: 143 °C (1013 hPa) Massefylde: 1,404 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008066 CAS-nummer: 461-58-5 EINECS: 207-312-8 Merck Index: 14,03092 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 209.0 to 213.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...