Cyanocobalamin ≥95.0% (by UV/Vis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

C0449-1G C0449-100MG
TCIAC0449-1GEA 706 DKK
TCIAC0449-1G TCIAC0449-100MG
Cyanocobalamin ≥95.0% (by UV/Vis)
Vitamin B12 (cyanocobalamin)
Formel: C₆₃H₈₈CoN₁₄O₁₄P
Molekylvægt: 1355.38 g/mol
Smeltepunkt: 320
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00151092
CAS-nummer: 68-19-9
EINECS: 200-680-0
Merck Index: 14,10014

Bestil nu

Specifikationer Testresultater

Appearance Red to Dark red to Dark purple powder to crystal
Purity (UV-vis method) 95.0 to 105.0 % (calcd.on dried substance)
abs-ratio (a/b) 0.53 to 0.59 (E278/E361)
abs-ratio (2a/2b) 0.29 to 0.32 (E550/E361)
Drying loss max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR