2-Amino-3-chlor-5-methylpyridin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Chloro-5-methyl-2-pyridylamine, 3-Chloro-5-methylpyridin-2-amine, 2-Amino-3-chloro-5-picoline

A1887-1G A1887-5G
TCIAA1887-1GEA 362 DKK
TCIAA1887-1G TCIAA1887-5G
2-Amino-3-chlor-5-methylpyridin ≥98.0% (by GC)
2-Amino-3-chloro-5-methylpyridine
Formel: C₆H₇ClN₂
MDL Number: MFCD07368858
CAS-nummer: 31430-41-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1b5f434d11ffcec7d75ad4b996ea71b324e65e8&vid=b1b5f434d11ffcec7d75ad4b996ea71b324e65e8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 62.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR