2,3-Dichlorphenylisocyanat ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

I0499-5G I0499-1G
TCIAI0499-5GEA 419 DKK
TCIAI0499-5G TCIAI0499-1G
2,3-Dichlorphenylisocyanat ≥97.0% (by GC)
2,3-Dichlorphenylisocyanat
Formel: C₇H₃Cl₂NO
Kogepunkt: 66°C
Massefylde: 1,42 g/cm³ (20 °C)
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00002000
CAS-nummer: 41195-90-8
EINECS: 255-253-1
UN nummer: 2250
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a62bbc2137d9b9140f91d2abb2900b430baa528&vid=8a62bbc2137d9b9140f91d2abb2900b430baa528&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR